5-Bromo-2,4-dihydroxybenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A18230-06 A18230-14
CAAAAA18230-06EA 63.67 CAD
CAAAAA18230-06 CAAAAA18230-14
5-Bromo-2,4-dihydroxybenzoic acid 98%
5-Bromo-2,4-dihydroxybenzoic acid

Purity: 98% CAS No.: 7355-22-8 Molecular Formula: C7H5BrO4 Formula Weight: 233.02 Melting Point: ca 205° dec. Beilstein Reference: 2967526

Formula: C₇H₅BrO₄
MW: 233.02 g/mol
Melting Pt: ∼ 205 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002452
CAS Number: 7355-22-8
EINECS: 230-881-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR