n-Tridecane 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18220-14
A18220-22
A18220-36
CAAAAA18220-14EA
67.62
CAD
CAAAAA18220-14
CAAAAA18220-22
CAAAAA18220-36
n-Tridecane 98+%
n-Tridecane
Purity: 98+%
CAS No.: 629-50-5
Molecular Formula: C13H28
Formula Weight: 184.37
Flash Point: 79°(174°F)
Refractive Index: 1.4250
Melting Point: -5°
Boiling Point: 235°
Beilstein Reference: 1733089
MDL No.: MFCD00008979
Formula:
H₃C(CH₂)₁₁CH₃ Boiling Pt: 235 °C Melting Pt: –5 °C Density: 0.755 Flash Pt: 79 °C(174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008979 CAS Number: 629-50-5 EINECS: 211-093-4 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...