4,4'-Bi-o-toluidine dihydrochloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: o-Toluidine dihydrochloride, o-Tolidine dihydrochloride

A18196-14 A18196-22
CAAAAA18196-14EA 80.74 CAD
CAAAAA18196-14 CAAAAA18196-22
4,4'-Bi-o-toluidine dihydrochloride 97%
4,4'-Bi-o-toluidine dihydrochloride

Purity: 97% CAS No.: 612-82-8 Molecular Formula: C14H16N2.2HCl Formula Weight: 285.22 Odor: Odorless Melting Point: 300° UN No.: 3077 MDL No.: MFCD00012960

Formula: C₁₄H₁₈Cl₂N₂
MW: 285.22 g/mol
Melting Pt: 130
Storage Temperature: Ambient
MDL Number: MFCD00012960
CAS Number: 612-82-8
EINECS: 210-322-5
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR