Orthophosphoric acid ≥85% in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18067-0D
A18067-0F
A18067-AP
CAAAAA18067-0DEA
718.9
CAD
CAAAAA18067-0D
CAAAAA18067-0F
CAAAAA18067-AP
Orthophosphoric acid ≥85% in aqueous solution
Orthophosphoric acid
Formula Weight: 98.00
Refractive Index: 1.4310
Melting Point: 19-21°
Merck Reference: 14,7344
Formula:
H₃PO₄ Melting Pt: 19…21 °C Density: 1.700 Storage Temperature: Ambient |
MDL Number:
MFCD00011340 CAS Number: 7664-38-2 EINECS: 231-633-2 UN: 1805 ADR: 8,III Merck Index: 14,7344 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...