Sodium sulfite, anhydrous 98%

Supplier: Thermo Scientific Chemicals

A17933-0C A17933-0E A17933-36
CAAAAA17933-0CEA 646.22 CAD
CAAAAA17933-0C CAAAAA17933-0E CAAAAA17933-36
Sodium sulfite, anhydrous 98%
Sodium sulphite

Purity: 98% CAS No.: 7757-83-7 Molecular Formula: Na2SO3 Formula Weight: 126.04 Melting Point: 500° dec. Merck Reference: 14,8682 MDL No.: MFCD00003503

Formula: Na₂SO₃
Melting Pt: >500 °C (dec.)
Density: 2.633
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS Number: 7757-83-7
EINECS: 231-821-4
REACH: 01-2119537420-49
Merck Index: 14,8682

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR