Lithium acetate dihydrate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17921-0B
A17921-30
CAAAAA17921-0BEA
412.38
CAD
CAAAAA17921-0B
CAAAAA17921-30
Lithium acetate dihydrate 99%
Lithium acetate dihydrate
Purity: 99%
CAS No.: 6108-17-4
Molecular Formula: LiOOCCH3.2H2O
Formula Weight: 102.01 (65.98anhy)
Melting Point: 54-57°
Beilstein Reference: 3564320
Merck Reference: 14,5521
MDL No.: MFCD00066949
Formula:
H₃CCOOLi·2H₂O Melting Pt: 54…57 °C Density: 1.300 Storage Temperature: Ambient |
MDL Number:
MFCD00066949 CAS Number: 6108-17-4 EINECS: 208-914-3 Merck Index: 14,5521 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...