Sodium thiosulfate pentahydrate 99+%
Supplier: Thermo Scientific Chemicals
A17914-0C
A17914-0E
A17914-36
CAAAAA17914-0CEA
450.3
CAD
CAAAAA17914-0C
CAAAAA17914-0E
CAAAAA17914-36
Sodium thiosulfate pentahydrate 99+%
Sodium thiosulphate pentahydrate
Purity: 99+%
CAS No.: 10102-17-7
Molecular Formula: Na2O3S2.5H2O
Formula Weight: 248.17 (158.10anhy)
Odor: Odorless
pH: 6.5-8.0
Melting Point: 48°
Boiling Point: 100° -5H2O
Merck Reference: 14,8694
MDL No.: MFCD00149186
Formula:
Na₂S₂O₃.5H₂O Boiling Pt: 100° …5H{2}O Melting Pt: 48 °C Density: 1.69 Storage Temperature: Ambient |
MDL Number:
MFCD00149186 CAS Number: 10102-17-7 EINECS: 231-867-5 Merck Index: 14,8694 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...