2,2'-Thiodiacetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A17880-22 A17880-36
CAAAAA17880-22EA 117.71 CAD
CAAAAA17880-22 CAAAAA17880-36
2,2'-Thiodiacetic acid 98%
2,2'-Thiodiacetic acid

Purity: 98% CAS No.: 123-93-3 Molecular Formula: C4H6O4S Formula Weight: 150.15 Melting Point: 127-131° Beilstein Reference: 1764392 Merck Reference: 14,9331 UN No.: 3261 MDL No.: MFCD00004359

Formula: C₄H₆O₄S
Boiling Pt: 168 °C (14 -rr)
Melting Pt: 127…131 °C
Density: 1.661 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004359
CAS Number: 123-93-3
EINECS: 204-663-9
UN: 3261
ADR: 8,III
Merck Index: 14,9331

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR