o-Tolidine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4,4'-Bi-o-toluidine

A17837-14 A17837-22
CAAAAA17837-14EA 140.62 CAD
CAAAAA17837-14 CAAAAA17837-22
o-Tolidine 98%
o-Tolidine

Purity: 98% CAS No.: 119-93-7 Molecular Formula: C14H16N2 Formula Weight: 212.30 Flash Point: 244°(471°F) Melting Point: 128-132° Boiling Point: 300-301° Merck Reference: 14,9514 UN No.: 3077 MDL No.: MFCD00014773

Formula: [-C₆H₃(CH₃)-₄-NH₂]₂
MW: 212.29 g/mol
Boiling Pt: 300…301 °C
Melting Pt: 128…132 °C
Density: ~ 1.234 g/cm³ (20 °C)
Flash Pt: 244 °C (471 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014773
CAS Number: 119-93-7
EINECS: 204-358-0
UN: 3077
ADR: 9,III
Merck Index: 14,9514

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR