4,7-Dimethyl-1,10-phenanthroline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17779-01
A17779-03
A17779-06
CAAAAA17779-01EA
76.95
CAD
CAAAAA17779-01
CAAAAA17779-03
CAAAAA17779-06
4,7-Dimethyl-1,10-phenanthroline 98%
4,7-Dimethyl-1,10-phenanthroline
Purity: 98%
CAS No.: 3248-05-3
Molecular Formula: C14H12N2
Formula Weight: 208.26
Melting Point: 193-195°
MDL No.: MFCD00004979
Formula:
C₁₄H₁₂N₂ Melting Pt: 193…195 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004979 CAS Number: 3248-05-3 EINECS: 221-827-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...