Bromothymol blue

Supplier: Thermo Scientific Chemicals

Synonyms: 3′,3″-Dibromothymolsulphonphthalein, 3′,3″-Dibromothymolsulfonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

A17746-09 A17746-18 A17746-30
CAAAAA17746-09EA 68.57 CAD
CAAAAA17746-09 CAAAAA17746-18 CAAAAA17746-30
Bromothymol blue
Bromothymol blue

CAS No.: 76-59-5 Molecular Formula: C27H28Br2O5S Formula Weight: 624.39 Melting Point: 200-202° Beilstein Reference: 373934 Merck Reference: 14,1445 MDL No.: MFCD00005872

Formula: C₂₇H₂₈Br₂O₅S
MW: 624.39 g/mol
Melting Pt: 200…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00005872
CAS Number: 76-59-5
EINECS: 200-971-2
Merck Index: 14,1445

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR