Ethyl (trimethylsilyl)acetate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17707-09
A17707-18
A17707-30
CAAAA17707-09EA
103.65
CAD
CAAAA17707-09
CAAAA17707-18
CAAAA17707-30
Ethyl (trimethylsilyl)acetate 98+%
Ethyl (trimethylsilyl)acetate
Purity: 98+%
CAS No.: 4071-88-9
Molecular Formula: (CH3)3SiCH2CO2CH2CH3
Formula Weight: 160.29
Flash Point: 35°(95°F)
Refractive Index: 1.4150
Boiling Point: 156-158°
Beilstein Reference: 1755902
UN No.: 3272
MDL No.: MFCD00009172
Formula:
C₇H₁₆O₂Si Boiling Pt: 156…158 °C Density: 0.876 Flash Pt: 35 °C(95 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009172 CAS Number: 4071-88-9 EINECS: 223-783-2 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...