Methyl-o-anisate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methoxybenzoic acid methyl ester, Methyl 2-methoxybenzoate, o-Anisic acid methyl ester, 2-Anisic acid methyl ester

A17703-14 A17703-22
CAAAA17703-14EA 53.88 CAD
CAAAA17703-14 CAAAA17703-22
Methyl-o-anisate 98+%
Methyl o-anisate

Purity: 98+% CAS No.: 606-45-1 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 110°(230°F) Refractive Index: 1.5340 Odor: Characteristic Boiling Point: 246-248° Beilstein Reference: 2207356 MDL No.: MFCD00008423

Formula: C₉H₁₀O₃
Boiling Pt: 246…248 °C
Density: 1.160
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008423
CAS Number: 606-45-1
EINECS: 210-118-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR