Ammonium sulfamate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ammonium sulfamidate, Ammonium sulphamidate, Ammonium amidosulfonate, Ammonium amidosulphonate
A17696-0B
A17696-30
CAAAAA17696-0BEA
194.34
CAD
CAAAAA17696-0B
CAAAAA17696-30
Ammonium sulfamate 98+%
Ammonium sulphamate
Purity: 98+%
CAS No.: 7773-06-0
Molecular Formula: H6N2O3S
Formula Weight: 114.12
Odor: Weak
pH: 5.2
Melting Point: 131-135°
Boiling Point: 160° dec.
Merck Reference: 14,554
MDL No.: MFCD00011429
Formula:
H₆N₂O₃S Boiling Pt: 160 °C (dec.) Melting Pt: 131…135 °C Density: Min. 1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011429 CAS Number: 7773-06-0 EINECS: 231-871-7 Merck Index: 14,554 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...