Dimethyltin dichloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dichlorodimethylstannane

A17661-06 A17661-14
CAAAAA17661-06EA 112.81 CAD
CAAAAA17661-06 CAAAAA17661-14
Dimethyltin dichloride 98%
Dimethyltin dichloride

Purity: 98% CAS No.: 753-73-1 Molecular Formula: (CH3)2SnCl2 Formula Weight: 219.69 Melting Point: 104-108° Boiling Point: 188-190° UN No.: 3146 MDL No.: MFCD00000517

Formula: C₂H₆Cl₂Sn
MW: 219.69 g/mol
Boiling Pt: 188…190 °C
Melting Pt: 104…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00000517
CAS Number: 753-73-1
EINECS: 212-039-2
UN: 2923
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR