Hexamethylditin 97%

Supplier: Thermo Scientific Chemicals
Danger

A17638-03 A17638-06 A17638-14
CAAAAA17638-03EA 93.54 CAD
CAAAAA17638-03 CAAAAA17638-06 CAAAAA17638-14
Hexamethylditin 97%
Hexamethylditin

Purity: 97% CAS No.: 661-69-8 Molecular Formula: C6H18Sn2 Formula Weight: 327.59 Storage Temperature: 2°C to 8°C Flash Point: 61°(142°F) Refractive Index: 1.5410 Odor: Characteristic Melting Point: 23° Boiling Point: 85°/45mm Beilstein Reference: 3600327 UN No.: 2788 MDL No.: MFCD00008277

Formula: C₆H₁₈Sn₂
Boiling Pt: 85 °C/45 mm
Melting Pt: 23 °C
Density: 1.58
Flash Pt: 61 °C (142 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00008277
CAS Number: 661-69-8
EINECS: 211-549-2
UN: 3146
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR