Sodium thiosulfate, anhydrous 99%

Supplier: Thermo Scientific Chemicals

A17629-0E A17629-36
CAAAAA17629-0EEA 262.28 CAD
CAAAAA17629-0E CAAAAA17629-36
Sodium thiosulfate, anhydrous 99%
Sodium thiosulphate

Purity: 99% CAS No.: 7772-98-7 Molecular Formula: Na2O3S2 Formula Weight: 158.10 Odor: Odorless Merck Reference: 14,8694 MDL No.: MFCD00003499

Formula: Na₂S₂O₃
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 48 °C
Density: 1.667
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5
Merck Index: 14,8694

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR