Manganese(II) sulfate monohydrate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Manganese sulphate monohydrate, Manganese sulfate monohydrate, Manganous sulfate monohydrate, Manganese(II) sulphate hydrate, Manganous sulphate monohydrate, Manganese(II) sulfate hydrate

A17615-0E A17615-22 A17615-36
CAAAAA17615-0EEA 502.44 CAD
CAAAAA17615-0E CAAAAA17615-22 CAAAAA17615-36
Manganese(II) sulfate monohydrate 99%
Manganese (II) sulphate monohydrate

Purity: 99% CAS No.: 10034-96-5 Molecular Formula: MnSO4.H2O Formula Weight: 169.02 (151.00anhy) Odor: Odorless pH: 3-3.5 Merck Reference: 14,5739 UN No.: 3077 MDL No.: MFCD00149159

Formula: MnSO₄·1H₂O
Melting Pt: 700 °C
Density: 2.95
Storage Temperature: Ambient
MDL Number: MFCD00149159
CAS Number: 10034-96-5
EINECS: 232-089-9
UN: 3077
ADR: 9,III
Merck Index: 14,5739

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR