Dithallium trioxide 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Thallium (III) oxide

A17601-09 A17601-18
CAAAAA17601-09EA 102.7 CAD
CAAAAA17601-09 CAAAAA17601-18
Dithallium trioxide 96%
Dithallium trioxide

Purity: 96% CAS No.: 1314-32-5 Molecular Formula: Tl2O3 Formula Weight: 456.74 Odor: Odorless Melting Point: 717° Boiling Point: 875° Merck Reference: 14,9268 UN No.: 1707 MDL No.: MFCD00011276

Formula: O₃Tl₂
MW: 456.76
Boiling Pt: 875 °C
Melting Pt: 717 °C
Density: 5.56
Storage Temperature: Ambient
MDL Number: MFCD00011276
CAS Number: 1314-32-5
EINECS: 215-229-3
UN: 1707
ADR: 6.1 II
Merck Index: 14,9268

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR