Triisopropyl borate 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tri-i-propylborate

A17592-0F A17592-AE A17592-AP
CAAAAA17592-0FEA 799.48 CAD
CAAAAA17592-0F CAAAAA17592-AE CAAAAA17592-AP
Triisopropyl borate 98+%
Triisopropyl borate

Purity: 98+% CAS No.: 5419-55-6 Molecular Formula: C9H21BO3 Formula Weight: 188.08 Flash Point: 17°(62°F) Refractive Index: 1.3760 Odor: Alcohol-like Melting Point: -59° Boiling Point: 139-141° Beilstein Reference: 1701469 UN No.: 2616 MDL No.: MFCD00008872

Formula: C₉H₂₁BO₃
MW: 188.07 g/mol
Boiling Pt: 139
Melting Pt: -59
Density: 0.815
Flash Pt: 17 °C (62 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008872
CAS Number: 5419-55-6
EINECS: 226-529-9
UN: 2616
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR