Tetrahexylammonium iodide 99%

Supplier: Thermo Scientific Chemicals
Warning

A17589-06 A17589-14
CAAAAA17589-06EA 67.62 CAD
CAAAAA17589-06 CAAAAA17589-14
Tetrahexylammonium iodide 99%
Tetrahexylammonium iodide

Purity: 99% CAS No.: 2138-24-1 Molecular Formula: [CH3(CH2)5]4NI Formula Weight: 481.59 Melting Point: 99-105° MDL No.: MFCD00041981

Formula: C₂₄H₅₂IN
Melting Pt: 99…105 °C
Density: 1,0922 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041981
CAS Number: 2138-24-1
EINECS: 218-382-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR