Tetrasodium diphosphate 98%

Supplier: Thermo Scientific Chemicals
Warning

A17546-0B A17546-30
CAAAAA17546-0BEA 139.51 CAD
CAAAAA17546-0B CAAAAA17546-30
Tetrasodium diphosphate 98%
tetra-Sodium diphosphate

Purity: 98% CAS No.: 7722-88-5 Molecular Formula: Na4O7P2 Formula Weight: 265.90 Melting Point: 988° Merck Reference: 14,9240 MDL No.: MFCD00003513

Formula: Na₄O₇P₂
Melting Pt: 988 °C
Density: 2.534
Storage Temperature: Ambient
MDL Number: MFCD00003513
CAS Number: 7722-88-5
EINECS: 231-767-1
Merck Index: 14,9240

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR