Murexide

Supplier: Thermo Scientific Chemicals

A17540-09 A17540-18 A17540-30
CAAAAA17540-09EA 94.82 CAD
CAAAAA17540-09 CAAAAA17540-18 CAAAAA17540-30
Murexide
Murexide

Indicator for calcium determinations


CAS No.: 3051-09-0 Molecular Formula: C8H8N6O6 Formula Weight: 284.19 Melting Point: 300° Beilstein Reference: 3582175 Merck Reference: 14,6306 MDL No.: MFCD00012777

Formula: C₈H₈N₆O₆
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00012777
CAS Number: 3051-09-0
EINECS: 221-266-6
Merck Index: 14,6306

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR