(Ethyl)triphenylphosphonium iodide 98+%

Supplier: Thermo Scientific Chemicals
Danger

A17533-14 A17533-22 A17533-36
CAAAAA17533-14EA 54.35 CAD
CAAAAA17533-14 CAAAAA17533-22 CAAAAA17533-36
(Ethyl)triphenylphosphonium iodide 98+%
(Ethyl)triphenylphosphonium iodide

Purity: 98+% CAS No.: 4736-60-1 Molecular Formula: CH3CH2P(C6H5)3I Formula Weight: 418.26 Melting Point: 164-169° Beilstein Reference: 3659323 UN No.: 2811 MDL No.: MFCD00040352

Formula: C₂₀H₂₀IP
Melting Pt: 164…169 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00040352
CAS Number: 4736-60-1
EINECS: 225-245-2
UN: 3464
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR