Diethyl phthalate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Phthalic acid diethyl ester
A17529-0B
A17529-0E
A17529-30
CAAAAA17529-0BEA
73.94
CAD
CAAAAA17529-0B
CAAAAA17529-0E
CAAAAA17529-30
Diethyl phthalate 99%
Diethyl phthalate
Purity: 99%
CAS No.: 84-66-2
Molecular Formula: C12H14O4
Formula Weight: 222.24
Flash Point: 160°(320°F)
Refractive Index: 1.5020
Melting Point: -64°
Boiling Point: 296-298°
Beilstein Reference: 1912500
Merck Reference: 14,7371
MDL No.: MFCD00009111
Formula:
C₁₂H₁₄O₄ MW: 222.24 g/mol Boiling Pt: 296…298 °C Melting Pt: –64 °C Density: 1.118 Flash Pt: 160 °C (320 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009111 CAS Number: 84-66-2 EINECS: 201-550-6 Merck Index: 14,7371 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...