DL-Isoleucine 99%

Supplier: Thermo Scientific Chemicals

A17521-14 A17521-22
CAAAAA17521-14EA 106.18 CAD
CAAAAA17521-14 CAAAAA17521-22
DL-Isoleucine 99%
DL-Isoleucine

Purity: 99% CAS No.: 443-79-8 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Melting Point: ca 280° dec. Beilstein Reference: 1721796 Merck Reference: 14,5179 MDL No.: MFCD00004268

Formula: C₆H₁₃NO₂
Melting Pt: ∼ 280 °C (dec.)
Density: 1.255 g/cm³ (-123.15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004268
CAS Number: 443-79-8
EINECS: 207-139-8
Merck Index: 14,5179

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR