Sodium chromate tetrahydrate 99+%

Supplier: Thermo Scientific Chemicals
Danger

A17499-22 A17499-36
CAAAAA17499-22EA 79.16 CAD
CAAAAA17499-22 CAAAAA17499-36
Sodium chromate tetrahydrate 99+%
Sodium chromate tetrahydrate

Purity: 99+% CAS No.: 10034-82-9 Molecular Formula: Na2CrO4.4H2O Formula Weight: 234.04 (161.97anhy) Odor: Odorless Merck Reference: 14,8601 MDL No.: MFCD00149167

Formula: CrNa₂O₄·4H₂O
MW: 234.03 g/mol
Melting Pt: 792.2 °C
Density: 2.73
Storage Temperature: Ambient
MDL Number: MFCD00149167
CAS Number: 10034-82-9
EINECS: 231-889-5
UN: 3288
ADR: 6.1,III
Merck Index: 14,8601

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR