Tetrabutylammonium tetrahydroborate 98%

Supplier: Thermo Scientific Chemicals
Danger

A17494-09 A17494-18 A17494-30
CAAAAA17494-09EA 126.4 CAD
CAAAAA17494-09 CAAAAA17494-18 CAAAAA17494-30
Tetrabutylammonium tetrahydroborate 98%
Tetrabutylammonium tetrahydroborate

Purity: 98% CAS No.: 33725-74-5 Molecular Formula: C16H40BN Formula Weight: 257.31 Flash Point: 140°(284°F) Melting Point: 125-131° UN No.: 3131 MDL No.: MFCD00012035

Formula: C₁₆H₄₀BN
Melting Pt: 125…131 °C
Flash Pt: 140 °C (284 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012035
CAS Number: 33725-74-5
EINECS: 251-658-2
UN: 3131
ADR: 4.3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR