Mercury(II) chloride 98+%

Supplier: Thermo Scientific Chemicals
Danger

A17472-0B A17472-18 A17472-30
CAAAAA17472-0BEA 625.68 CAD
CAAAAA17472-0B CAAAAA17472-18 CAAAAA17472-30
Mercury(II) chloride 98+%
Mercury (II) chloride

Purity: 98+% CAS No.: 7487-94-7 Molecular Formula: HgCl2 Formula Weight: 271.50 Refractive Index: 1.859 Odor: Odorless Boiling Point: 302° Merck Reference: 14,5876 UN No.: 1624 MDL No.: MFCD00011041

Formula: HgCl₂
Boiling Pt: 302 °C
Melting Pt: 280…282 °C
Density: 5.4
Storage Temperature: Ambient
MDL Number: MFCD00011041
CAS Number: 7487-94-7
EINECS: 231-299-8
UN: 1624
ADR: 6.1,II
Merck Index: 14,5876

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR