Tetramethylammonium hexafluorophosphate 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetramethylammoniumhexafluorophosphate

A17467-14 A17467-22
CAAAAA17467-14EA 251.22 CAD
CAAAAA17467-14 CAAAAA17467-22 CAAAAA17467-36
Tetramethylammonium hexafluorophosphate 99+%
Tetramethylammonium hexafluorophosphate

Purity: 99+% CAS No.: 558-32-7 Molecular Formula: C4H12F6NP Formula Weight: 219.10 Odor: Odorless Melting Point: 300° Beilstein Reference: 3730394 UN No.: 2923 MDL No.: MFCD00012012

Formula: C₄H₁₂F₆NP
Melting Pt: >300 °C
Density: 1.617 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012012
CAS Number: 558-32-7
EINECS: 209-194-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR