Potassium trifluoroacetate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Trifluoroacetic acid potassium salt, Potassium 2,2,2-trifluoroacetate
A17398-14
A17398-22
CAAAAA17398-14EA
81.53
CAD
CAAAAA17398-14
CAAAAA17398-22
Potassium trifluoroacetate 98%
Potassium trifluoroacetate
Purity: 98%
CAS No.: 2923-16-2
Molecular Formula: CF3CO2K
Formula Weight: 152.12
pH: 7
Melting Point: 142-145°
Beilstein Reference: 3717603
UN No.: 2811
MDL No.: MFCD00013215
Formula:
C₂F₃KO₂ Melting Pt: 142…145 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013215 CAS Number: 2923-16-2 EINECS: 220-877-5 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...