Terbium triacetate hydrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Terbium(III) acetate hydrate

A17388-06 A17388-14
CAAAA17388-06EA 257.54 CAD
CAAAA17388-06 CAAAA17388-14
Terbium triacetate hydrate 99%
Terbium triacetate hydrate

Purity: 99.99% CAS No.: 100587-92-6 Molecular Formula: Tb(OOCCH3)3.xH2O Formula Weight: 336.08(anhy) MDL No.: MFCD00149233

Formula: C₆H₉O₆Tb·H₂O
MW: 354.07
Storage Temperature: Ambient
MDL Number: MFCD00149233
CAS Number: 100587-92-6
EINECS: 247-065-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR