Sodium metabisulfite 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17351-0C
A17351-0E
A17351-36
CAAAAA17351-0CEA
322.32
CAD
CAAAAA17351-0C
CAAAAA17351-0E
CAAAAA17351-36
Sodium metabisulfite 97%
Sodium metabisulphite
Purity: 97%
CAS No.: 7681-57-4
Molecular Formula: Na2O5S2
Formula Weight: 190.10
Odor: Like rotten eggs
pH: 3.5-5.0
Melting Point:
Formula:
Na₂S₂O₅ Melting Pt: >300 °C Density: 1.480 Storage Temperature: Ambient |
MDL Number:
MFCD00167602 CAS Number: 7681-57-4 EINECS: 231-673-0 REACH: 01-2119531326-45 Merck Index: 14,8638 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...