4-Picoline N-oxide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Methylpyridine 1-oxide, 4-Methylpyridine-N-oxide, 4-Methylpyridine-1-oxide, 4-Picoline N-oxide
A17330-14
A17330-22
CAAAAA17330-14EA
68.57
CAD
CAAAAA17330-14
CAAAAA17330-22
4-Picoline N-oxide 98%
4-Picoline N-oxide
Purity: 98%
CAS No.: 1003-67-4
Molecular Formula: C6H7NO
Formula Weight: 109.13
Flash Point: 149°(300°F)
Melting Point: 182-185°
Boiling Point: 152°/11mm
Beilstein Reference: 106329
MDL No.: MFCD00006210
Formula:
C₆H₇NO Boiling Pt: 152 °C/11 mm Melting Pt: 182…185 °C Density: 1,01…1.02 g/cm³ (185 °C) Flash Pt: 149 °C(300 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006210 CAS Number: 1003-67-4 EINECS: 213-712-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...