L(+)-Lactic acid ∼85 - 90% in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

A17268-0E A17268-22 A17268-36
CAAAAA17268-0EEA 390.26 CAD
CAAAAA17268-0E CAAAAA17268-22 CAAAAA17268-36
L(+)-Lactic acid ∼85 - 90% in aqueous solution
L(+)-Lactic acid

CAS No.: 79-33-4 Molecular Formula: C3H6O3 Formula Weight: 90.08 Flash Point: 110°(230°F) Refractive Index: 1.4270 Odor: Odorless pH: 2.8 Melting Point: 18° Beilstein Reference: 1720251 Merck Reference: 14,5337 Optical Rotation: -13° (c=2.5 in 1N NaOH) MDL No.: MFCD00064266

Formula: C₃H₆O₃
Melting Pt: 18 °C
Density: 1.209
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064266
CAS Number: 79-33-4
EINECS: 201-196-2
Merck Index: 14,5337

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR