O-Dianisidine dihydrochloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17175-03
A17175-06
CAAAA17175-03EA
71.42
CAD
CAAAA17175-03
CAAAA17175-06
O-Dianisidine dihydrochloride 99%
o-Dianisidine dihydrochloride
Purity: 99%
CAS No.: 20325-40-0
Molecular Formula: C14H16N2O2.2HCl
Formula Weight: 317.21
Melting Point: 268° dec.
Beilstein Reference: 3917996
Formula:
C₁₄H₁₈Cl₂N₂O₂ Melting Pt: 268 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012488 CAS Number: 20325-40-0 EINECS: 243-737-5 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...