O-Dianisidine 98+%

Supplier: Thermo Scientific Chemicals
Danger

A17150-06 A17150-14 A17150-22
CAAAA17150-06EA 61.78 CAD
CAAAA17150-06 CAAAA17150-14 CAAAA17150-22
O-Dianisidine 98+%
o-Dianisidine

Purity: 98+% CAS No.: 119-90-4 Molecular Formula: C14H16N2O2 Formula Weight: 244.30 Flash Point: 206°(402°F) Melting Point: 134-138° Beilstein Reference: 1879884 Merck Reference: 14,2991 MDL No.: MFCD00008372

Formula: C₁₄H₁₆N₂O₂
Melting Pt: 134…138 °C
Flash Pt: 206 °C(402 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008372
CAS Number: 119-90-4
EINECS: 204-355-4
Merck Index: 14,2991

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR