Phenolphthalein 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

A17135-0B A17135-18 A17135-30
CAAAAA17135-0BEA 301.78 CAD
CAAAAA17135-0B CAAAAA17135-18 CAAAAA17135-30
Phenolphthalein 98%
Phenolphthalein

Purity: 98% CAS No.: 77-09-8 Molecular Formula: C20H14O4 Formula Weight: 318.33 Melting Point: 258-262° Beilstein Reference: 284423 Merck Reference: 14,7243 MDL No.: MFCD00005913

Formula: C₂₀H₁₄O₄
MW: 318.33 g/mol
Melting Pt: 258…262 °C
Density: 1.299
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
EINECS: 201-004-7
REACH: 01-2119498295-24
Merck Index: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR