α-Ionone 90+%, Technical Grade

Supplier: Thermo Scientific Chemicals

Synonyms: (E)-4-(2,6,6-Trimethylcyclohex-2-en-1-yl)but-3-en-2-one

A17098-22 A17098-36
CAAAAA17098-22EA 93.54 CAD
CAAAAA17098-22 CAAAAA17098-36
α-Ionone 90+%, Technical Grade
α-Ionone

Purity: 90+% CAS No.: 127-41-3 Molecular Formula: C13H20O Formula Weight: 192.30 Flash Point: 117°(243°F) Refractive Index: 1.4980 Boiling Point: 237° Beilstein Reference: 2046084 Merck Reference: 14,5056 MDL No.: MFCD00001565

Formula: C₁₃H₂₀O
MW: 192.3 g/mol
Boiling Pt: 237 °C
Density: 0.931
Flash Pt: 117 °C(243 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001565
CAS Number: 127-41-3
EINECS: 204-841-6
Merck Index: 14,5056

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR