Sodium salicylate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium 2-hydroxybenzoate, 2-Hydroxybenzoic acid sodium salt, Salicylic acid sodium salt

A17056-0B A17056-30
CAAAAA17056-0BEA 248.06 CAD
CAAAAA17056-0B CAAAAA17056-30
Sodium salicylate 99%
Sodium salicylate

Purity: 99% CAS No.: 54-21-7 Molecular Formula: C7H5NaO3 Formula Weight: 160.11 Odor: Odorless Melting Point: 300° Beilstein Reference: 3732792 Merck Reference: 14,8332 MDL No.: MFCD00002440

Formula: HOC₆H₄COONa
MW: 160.10 g/mol
Melting Pt: >300 °C
Density: 0.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002440
CAS Number: 54-21-7
EINECS: 200-198-0
Merck Index: 14,8332

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR