Tannic acid
Supplier: Thermo Scientific Chemicals
A17022-0B
A17022-30
CAAAAA17022-0BEA
281.24
CAD
CAAAAA17022-0B
CAAAAA17022-30
Tannic acid
Tannic acid
Used for enzyme immobilization and protein adsorption
CAS No.: 1401-55-4
Molecular Formula: C76H52O46
Formula Weight: 1701.23
Odor: Odorless
pH: 3.5
Melting Point: ca 218° dec.
Beilstein Reference: 8186396
Merck Reference: 14,9052
MDL No.: MFCD00066397
Formula:
C₇₆H₅₂O₄₆ Melting Pt: ∼ 218 °C (dec.) Density: Min. 1 g/cm³ (20 °C) Flash Pt: 199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066397 CAS Number: 1401-55-4 EINECS: 215-753-2 Merck Index: 14,9052 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...