Adenine sulfate 98+%

Supplier: Thermo Scientific Chemicals
Warning

A16964-09 A16964-18
CAAAA16964-09EA 79.16 CAD
CAAAA16964-09 CAAAA16964-18
Adenine sulfate 98+%
Adenine sulphate

Purity: 98+% CAS No.: 321-30-2 Molecular Formula: C10H10N10.H2SO4 Formula Weight: 368.34 Odor: Odorless Melting Point: ca 210° dec. Beilstein Reference: 3820263 Merck Reference: 14,152 MDL No.: MFCD00213655

Formula: C₁₀H₁₂N₁₀O₄S
Melting Pt: ca 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00213655
CAS Number: 321-30-2
EINECS: 206-286-5
Merck Index: 14,152

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR