Potassium hexacyanoferrate(III) 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: Potassium ferricyanide(III), Tripotassium hexacyanoferrate, Potassium ferricyanide

A16946-0B A16946-0I A16946-30
CAAAAA16946-0BEA 169.06 CAD
CAAAAA16946-0B CAAAAA16946-0I CAAAAA16946-30
Potassium hexacyanoferrate(III) 98+%
Potassium hexacyanoferrate (III)

Purity: 98+% CAS No.: 13746-66-2 Molecular Formula: C6FeK3N6 Formula Weight: 329.26 Melting Point: dec. Merck Reference: 14,7630 MDL No.: MFCD00011392

Formula: K₃Fe(CN)₆
MW: 329.25 g/mol
Melting Pt: Dec.
Density: 1.89
Storage Temperature: Ambient
MDL Number: MFCD00011392
CAS Number: 13746-66-2
EINECS: 237-323-3
Merck Index: 14,7630

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR