3-Chloro-1-propanol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16871-14
A16871-22
A16871-36
CAAAAA16871-14EA
54.04
CAD
CAAAAA16871-14
CAAAAA16871-22
CAAAAA16871-36
3-Chloro-1-propanol 98%
3-Chloro-1-propanol
Purity: 98%
CAS No.: 627-30-5
Molecular Formula: ClCH2CH2CH2OH
Formula Weight: 94.54
Flash Point: 73°(163°F)
Refractive Index: 1.4450
Odor: Pleasant
pH: 3-4
Melting Point: -20°
Boiling Point: 160-162°
Beilstein Reference: 773655
UN No.: 2849
MDL No.: MFCD00002943
Formula:
C₃H₇ClO Boiling Pt: 160…162 °C Melting Pt: –20 °C Density: 1.131 Flash Pt: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002943 CAS Number: 627-30-5 EINECS: 210-992-9 UN: 2849 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...