2-Ethylphenylhydrazine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Danger

A16853-18 A16853-30
CAAAA16853-18EA 336.54 CAD
CAAAA16853-18 CAAAA16853-30
2-Ethylphenylhydrazine hydrochloride 98%
2-Ethylphenylhydrazine hydrochloride

Purity: 98% CAS No.: 19398-06-2 Molecular Formula: C8H12N2.HCl Formula Weight: 172.66 Melting Point: ca 178° dec. Beilstein Reference: 3697547 UN No.: 3077 MDL No.: MFCD00071599

Formula: C₈H₁₃ClN₂
Melting Pt: ca 178 °C (dec.)
Density: 1.21
Storage Temperature: Ambient
MDL Number: MFCD00071599
CAS Number: 19398-06-2
EINECS: 421-460-2
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR