L-Threonine 98+%
Supplier: Thermo Scientific Chemicals
A16851-14
A16851-22
A16851-36
CAAAA16851-14EA
90.69
CAD
CAAAA16851-14
CAAAA16851-22
CAAAA16851-36
L-Threonine 98+%
L-Threonine
Purity: 98+%
CAS No.: 72-19-5
Molecular Formula: C4H9NO3
Formula Weight: 119.12
Melting Point: 255° dec.
Beilstein Reference: 1721646
Merck Reference: 14,9380
Optical Rotation: -28° (c=5 in water)
MDL No.: MFCD00064270
Formula:
C₄H₉NO₃ Melting Pt: 255 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064270 CAS Number: 72-19-5 EINECS: 200-774-1 Merck Index: 14,9380 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...