Tetrabutylphosphonium iodide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16792-06
A16792-14
CAAAAA16792-06EA
89.9
CAD
CAAAAA16792-06
CAAAAA16792-14
CAAAAA16792-22
Tetrabutylphosphonium iodide 98%
Tetrabutylphosphonium iodide
Purity: 98%
CAS No.: 3115-66-0
Molecular Formula: C16H36IP
Formula Weight: 386.34
Melting Point: 98-100°
MDL No.: MFCD00050246
Formula:
C₁₆H₃₆IP MW: 386.34 g/mol Melting Pt: 98…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00050246 CAS Number: 3115-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...