Cesium sulfate 99%

Supplier: Thermo Scientific Chemicals
Warning

A16767-14 A16767-22 A16767-36
CAAAAA16767-14EA 88.64 CAD
CAAAAA16767-14 CAAAAA16767-22 CAAAAA16767-36
Cesium sulfate 99%
Cesium sulphate

Purity: 99% CAS No.: 10294-54-9 Molecular Formula: Cs2SO4 Formula Weight: 361.87 Odor: Odorless Melting Point: 1010° MDL No.: MFCD00010959

Formula: Cs₂O₄S
Boiling Pt: 1627 °C (1013 hPa)
Melting Pt: 1010 °C
Density: 4.24
Storage Temperature: Ambient
MDL Number: MFCD00010959
CAS Number: 10294-54-9
EINECS: 233-662-6
Merck Index: 14,2016

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR