3'-Bromopropiophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

A16757-06 A16757-14
CAAAAA16757-06EA 47.56 CAD
CAAAAA16757-06 CAAAAA16757-14
3'-Bromopropiophenone 97%
3'-Bromopropiophenone

Purity: 97% CAS No.: 19829-31-3 Molecular Formula: C9H9BrO Formula Weight: 213.08 Flash Point: 110°(230°F) Melting Point: 38-41° Boiling Point: 144-145°/20mm Beilstein Reference: 1936736 MDL No.: MFCD00000084

Formula: C₉H₉BrO
Boiling Pt: 144…145 °C/20 mm
Melting Pt: 38…41 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000084
CAS Number: 19829-31-3
EINECS: 243-353-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR