Trichloro(phenyl)silane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Phenyltrichlorosilane
A16713-22
A16713-36
CAAAAA16713-22EA
51.35
CAD
CAAAAA16713-22
CAAAAA16713-36
Trichloro(phenyl)silane 97%
Trichloro(phenyl)silane
Purity: 97%
CAS No.: 98-13-5
Molecular Formula: C6H5Cl3Si
Formula Weight: 211.55
Flash Point: 91°(195°F)
Refractive Index: 1.5230
Odor: Odorless
Melting Point: -127°
Boiling Point: 201°
Beilstein Reference: 508730
UN No.: 1804
MDL No.: MFCD00000479
Formula:
C₆H₅Cl₃Si MW: 211.55 g/mol Boiling Pt: 201 °C Melting Pt: –127 °C Density: 1.324 Flash Pt: 91 °C (195 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000479 CAS Number: 98-13-5 EINECS: 202-640-8 UN: 1804 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...